Orders LinkContactUsLinkComponent
Testing Familial Relationships Using Simulated Lab Activity
Testing Familial Relationships Using Simulated Lab Activity
Testing Familial Relationships Using Simulated Blood Refill Kit, 6
Catalog # 470023-690
Supplier:   Avantor
Testing Familial Relationships Using Simulated Lab Activity
Catalog # 470023-690
Supplier:  Avantor
Testing Familial Relationships Using Simulated Lab Activity
Testing Familial Relationships Using Simulated Lab Activity
Testing Familial Relationships Using Simulated Blood Refill Kit, 6
Price
$873.00
Log in Para disponibilidad
Kit de 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Students determine what child belongs to which set of parents.
  • Kit Employs NGSS Life Science DCIs LS1 and LS3
  • Pre and Post-Laboratory Assessments Included
  • Vocabulary Words and Assessments Target Common Core Standards
  • Estimated Lab Time: 30 Minutes
  • Kit Aligned with National Standards
Students use safe and non-toxic Simulated Blood to determine the blood types of two set of parents and two children and ascertain which child belongs to which family. During this activity, students employ the basic Framework Principles of Structures and Properties (LS1) and Heredity (LS3). This activity can serve as an introduction to forensic science applications as well. This activity helps students to understand how familial relationships can be determined using blood typing.
This kit aligns with all published National Standards. Pre and post-laboratory assessments and vocabulary words target specific Science and Engineering Practices and Common Core standards. The teacher guide also contains specific "Did You Know" concepts that link the real world with the scientific standards explored. The kit includes a student copymaster and material for six setups.
Ordering information: This kit includes Simulated Blood: Mr. Smith, Mrs. Smith, Mr. Jones, Mrs. Jones, Child One, and Child Two. It also includes simulated anti-A Serum, simulated anti-B Serum, blood-typing slides, and toothpicks.
Especificaciones
  • Description:
    Testing Familial Relationships Using Simulated Blood Refill Kit
  • No. of Lab Groups:
    6
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 