Orders LinkContactUsLinkComponent
Plastic Vials
Plastic Vials
Vials, without caps, No
Catalog # 470020-894
Plastic Vials
Catalog # 470020-894
Supplier:  
Plastic Vials
Plastic Vials
Vials, without caps, No
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
PLASTIC VIALS W/CAPS PKG/10 Catalog #470020-892
El producto 470020-892 es similar a 470020-894, pero el fabricante y las especificaciones pueden variar.
Product Details & Documents
Made of safe, unbreakable plastic, the vials are available with or without friction-fit caps to prevent escape.
  • 3 oz. capacity
Use our foam plugs for ventilation. Size: 4×1 ³/₈".
Caution: Note: Not suitable for holding liquid.
Especificaciones
  • Cap included:
    No
  • Description:
    Vials, without caps