Orders LinkContactUsLinkComponent
Ceramic Fiber Squares
Ceramic Fiber Squares
Ceramic Fiber Squares, 15 x 15 cm
Catalog # 470122-538
Supplier #:  550F-6X6
Ceramic Fiber Squares
Catalog # 470122-538
Supplier:  
Ceramic Fiber Squares
Ceramic Fiber Squares
Ceramic Fiber Squares, 15 x 15 cm
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
CERAMIC FIBER SQ - 5X5IN PK12 Catalog #470122-536
Product 470122-536 is a functional alternative to 470122-538 but Packaging, Quantity, Manufacturer and/or specifications may vary.
Product Details & Documents
Made of ceramic fiber with organic binder, for use under hot vessels to protect table tops. Does not contain asbestos. Package of 12.
Specifications
  • Description:
    Ceramic Fiber Squares, 15 x 15 cm