Orders LinkContactUsLinkComponent
Ceramic Fiber Squares
Ceramic Fiber Squares
Ceramic Fiber Squares, 13 x 13 cm
Catalog # 470122-536
Supplier:   ARMIL/CFS INC SE
Número de proveedor #:  550F-5X5
Ceramic Fiber Squares
Catalog # 470122-536
Supplier:  ARMIL/CFS INC SE
Ceramic Fiber Squares
Ceramic Fiber Squares
Ceramic Fiber Squares, 13 x 13 cm
Price
$1,124.64
Log in Para disponibilidad
Paquete de 12
Product Details & Documents
Made of ceramic fiber with organic binder, for use under hot vessels to protect table tops. Does not contain asbestos. Package of 12.
Especificaciones
  • Description:
    Ceramic Fiber Squares, 13 x 13 cm