Ceramic Fiber Squares
Ceramic Fiber Squares, 13 x 13 cm
Catalog # 470122-536
Supplier: ARMIL/CFS INC SE
Número de proveedor #: 550F-5X5
Jump to:
Product Details & Documents
Made of ceramic fiber with organic binder, for use under hot vessels to protect table tops. Does not contain asbestos. Package of 12.
Especificaciones
- Description:Ceramic Fiber Squares, 13 x 13 cm