Soil Thermometer
Thermometer, Soil, Stainless Steel, 0...80 °C, 1 °C
Catalog # 470106-366
Supplier: TAYLOR PRECISION PRODUCTS SE
Número de proveedor #: 5976N
Jump to:
Product Details & Documents
Determine Soil Temperature Quickly.
- Large visible display of optimal growing conditions
- Durable
- Aligned with GLOBE protocols
Use this stainless steel thermometer to test soil temperatures for planting. The dial, which is 6 cm in diameter, records temperatures from 0 to 80 °C and 20 to 180 °F. The 12.5 cm stem is long enough for most planting depths.
Especificaciones
- Description:Thermometer, Soil, Stainless Steel
- Divisions:1 °C
- Range:0...80 °C