Orders LinkContactUsLinkComponent
Burner Tripods
Burner Tripods
Tripod, Burner, 23 cm x 11 cm x 9 cm, —
Catalog # 470104-578
Número de proveedor #:  TRS5X8-S
Burner Tripods
Catalog # 470104-578
Burner Tripods
Burner Tripods
Tripod, Burner, 23 cm x 11 cm x 9 cm, —
Price
$298.49
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Good quality, enameled cast iron tripods for supporting small objects over a burner.
  • Cast iron, enameled finish
  • Removable zinc plated legs
Good quality, enameled cast iron tripods for supporting small objects over a burner. The legs detach for easy storage.
Especificaciones
  • Ext.Ø:
  • Description:
    Tripod, Burner, 23 cm x 11 cm x 9 cm