Curved Microdissection Forceps
Straight Microdissection Forceps, Stainless steel, 115 mm (4.5"), Straight
Catalog # 470007-556
Supplier: DUNRITE INSTRUMENTS SE
Número de proveedor #: 6853305
Jump to:
Product Details & Documents
Micro fine, curved or straight points enhance control when probing the most delicate structures.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Especificaciones
- Description:Straight Microdissection Forceps
- Length:115 mm (4.5")
- Material:Stainless steel
- Tip Style:Straight