Orders LinkContactUsLinkComponent
Hook Collars
Hook Collar, Fits 19 mm (0.75") Rod
  470006-702
 :  HKCL02
Hook Collars
  470006-702
Hook Collars
Hook Collars
Hook Collar, Fits 19 mm (0.75") Rod
$87.42
Each (1UN)
Quickly add a hook to a support rod.
  • Plated steel
  • Thumbscrew
Plated steel collar clamps feature thumbscrews and hooks for suspending pulleys, balances, etc. from support rods or crossbars.
  • Description:
    Hook Collar, Fits 19 mm (0.75") Rod