Ward's® Slate (Dark Gray)
Slate (Dark Gray), student specimens, 1×1 - 1×1"
470026-282
: Avantor
Chlorite-mica with perfect slaty cleavage.
- Collected in Vermont
- Multiple sizes and pack options to fit specific needs
- Cut by Ward's geologists to guarantee top market quality
- Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All Specimen dimensions are considered an average size range.
- Description:Slate (Dark Gray), student specimens
- Specimen Dimensions:1×1 - 1×1"