Orders LinkContactUsLinkComponent
Ward's® Shale (Argillaceous)
Ward's® Shale (Argillaceous)
Shale (Argillaceous), student specimens, 1×1 - 1×1"
Catalog # 470026-256
Supplier:   Avantor
Ward's® Shale (Argillaceous)
Catalog # 470026-256
Supplier:  Avantor
Ward's® Shale (Argillaceous)
Ward's® Shale (Argillaceous)
Shale (Argillaceous), student specimens, 1×1 - 1×1"
Price
$227.95
Log in Para disponibilidad
Paquete de 10
Product Details & Documents
Typical green-gray, fissile, clay-rich.
  • Collected in New York
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
All specimen dimensions are considered an average size range.
Especificaciones
  • Description:
    Shale (Argillaceous), student specimens
  • Specimen Dimensions:
    1×1 - 1×1"