Ward's® Sandstone (Red)
Sandstone (Red), bulk pack, 1 kg, Size will vary
470026-214
: Avantor
Hematite-cemented quartz; orthoquartzite.
- Collected in New York
- Multiple sizes and pack options to fit specific needs
- Cut by Ward's geologists to guarantee top market quality
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All Specimen dimensions are considered an average size range.
- Description:Sandstone (Red), bulk pack, 1 kg
- Specimen Dimensions:Size will vary