Orders LinkContactUsLinkComponent
Ward's® Sandstone (Red)
Ward's® Sandstone (Red)
Sandstone (Red), student specimens, 1×1 - 1×1"
Catalog # 470026-212
Supplier:   Avantor
Ward's® Sandstone (Red)
Catalog # 470026-212
Supplier:  Avantor
Ward's® Sandstone (Red)
Ward's® Sandstone (Red)
Sandstone (Red), student specimens, 1×1 - 1×1"
Price
$246.28
Log in Para disponibilidad
Paquete de 10
Product Details & Documents
Hematite-cemented quartz; orthoquartzite.
  • Collected in New York
  • Multiple sizes and pack options to fit specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All Specimen dimensions are considered an average size range.
Especificaciones
  • Description:
    Sandstone (Red), student specimens
  • Specimen Dimensions:
    1×1 - 1×1"