Ward's® Limestone (Encrinal)
Limestone (Encrinal), Bulk Pack, 1 kg, Size will vary
470026-088
: Avantor
Gray with line crinoid columnal fragments.
Collected in Ohio
- Multiple sizes and pack options to fit your specific needs
- Cut by Ward's geologists to guarantee top market quality
- Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kg of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range. Some crinoids may require a hand lens to observe.
- Description:Limestone (Encrinal), Bulk Pack, 1 kg
- Specimen Dimensions:Size will vary