Orders LinkContactUsLinkComponent
Limestone (Coquina)
Limestone (Coquina)
Limestone (Coquina), Hand Specimen, 2×3" - 3×4"
Catalog # 470026-078
Supplier:   Avantor
Limestone (Coquina)
Catalog # 470026-078
Supplier:  Avantor
Limestone (Coquina)
Limestone (Coquina)
Limestone (Coquina), Hand Specimen, 2×3" - 3×4"
Price
$278.90
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Buff-colored, coarse, detrital limestone, with cemented shell fragments.
  • Collected in Florida
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Especificaciones
  • Description:
    Limestone (Coquina), Hand Specimen
  • Specimen Dimensions:
    2×3" - 3×4"