Ward's® Anorthite
Product Details & Documents
Feldspar group; calcium end member plagioclase crystals in pyroxene matrix.
Collected in California
- Multiple sizes and pack options to fit your specific needs
- Cut by Ward's Geologists to guarantee top market quality
- Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25 to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Especificaciones
- Description:Anorthite, hand specimen, each
- Specimen Dimensions:2×3 - 3×4"