Orders LinkContactUsLinkComponent
Ward's® Igneous Rock Collection
Ward's® Igneous Rock Collection
Igneous Rock Collection, 12
Catalog # 470025-222
Supplier:   Avantor
Ward's® Igneous Rock Collection
Catalog # 470025-222
Supplier:  Avantor
Ward's® Igneous Rock Collection
Ward's® Igneous Rock Collection
Igneous Rock Collection, 12
Price
$884.40
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Samples of Intrusive and Extrusive Igneous Rocks
  • Introduce concepts of the Earth's crust
  • Compartmented box
Provide your students with an excellent introduction to the forces at work beneath the Earth’s crust. The set is housed in a compartmented collection box and includes an identification list.
Contents:
12 numbered samples, 1" x 2"
Rocks:
basalt, diorite, gabbro, granite, obsidian, pegmatite, peridotite, pumice, rhyolite (2), scoria, syenite
Hand Specimen Set:
Large hand samples are ideal for demonstration and display. This set is housed in individual, labeled plastic display trays and comes with an identification list.
Contents:
12 numbered samples, 3" x 4"
Documents
Especificaciones
  • Description:
    Igneous Rock Collection
  • No. of Samples:
    12