Ward's® Labradorite (Cleavage)
Labradorite Thin Section, —
Catalog # 470024-946
Supplier: Avantor
Jump to:
Product Details & Documents
Feldspar group; good cleavages of pure soda lime feldspar; most show twinning and labradorescence.
- Collected in Quebec
- Multiple sizes and pack options to fit your specific needs
- Cut by Ward's geologists to guarantee top market quality
- Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Especificaciones
- Description:Labradorite Thin Section
- Specimen Dimensions:—