Orders LinkContactUsLinkComponent
Shale (Carbonaceous)
Shale (Carbonaceous)
Shale (Carbonaceous), Student Specimens, 1×1 - 1×1"
Catalog # 470015-512
Supplier:   Avantor
Shale (Carbonaceous)
Catalog # 470015-512
Supplier:  Avantor
Shale (Carbonaceous)
Shale (Carbonaceous)
Shale (Carbonaceous), Student Specimens, 1×1 - 1×1"
Price
$227.38
Log in Para disponibilidad
Paquete de 10
Product Details & Documents
Black, fissile; commonly with carbonized fossil plant impressions.
  • Collected in Pennsylvania
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kg of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Especificaciones
  • Description:
    Shale (Carbonaceous), Student Specimens
  • Specimen Dimensions:
    1×1 - 1×1"