Orders LinkContactUsLinkComponent
Ward's® Pegmatite
Ward's® Pegmatite
Pegmatite, student specimens, 1×1 - 1×1"
Catalog # 470015-470
Supplier:   Avantor
Ward's® Pegmatite
Catalog # 470015-470
Supplier:  Avantor
Ward's® Pegmatite
Ward's® Pegmatite
Pegmatite, student specimens, 1×1 - 1×1"
Price
$222.20
Log in Para disponibilidad
Paquete de 10
Product Details & Documents
Classic, very coarse-grained igneous texture.
  • Collected in South Dakotah
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Especificaciones
  • Description:
    Pegmatite, student specimens
  • Specimen Dimensions:
    1×1 - 1×1"