Orders LinkContactUsLinkComponent
Ward's® Coal (Lignite)
Ward's® Coal (Lignite)
Coal (Lignite), student specimens, 1×1" - 1×1"
  470015-464
 :   Avantor
Ward's® Coal (Lignite)
  470015-464
 :  Avantor
Ward's® Coal (Lignite)
Ward's® Coal (Lignite)
Coal (Lignite), student specimens, 1×1" - 1×1"
$242.85
Paquete de 10
Brownish black, intermediate stage between peat and bituminous coal.
  • Collected in North Dakota
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student Specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
  • Description:
    Coal (Lignite), student specimens
  • Specimen Dimensions:
    1×1" - 1×1"