Orders LinkContactUsLinkComponent
Utility Pails
Utility Pails
Utility Pail with Lid, 19 L (5 gal.)
Catalog # 470006-568
Supplier:   Avantor
Utility Pails
Catalog # 470006-568
Supplier:  Avantor
Utility Pails
Utility Pail with Lid, 19 L (5 gal.)
Price
$445.60
Log in for availability
Each (1UN)
Product Details & Documents
PE utility pails with lid.
Specifications
  • Capacity:
    19 L (5 gal.)
  • Description:
    Utility Pail with Lid
Accessories
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • Tagboard
    Tagboard
    470149-944
    23×30.5 cm (9×12")
    Log in for availability
    Paquete de 1 - $176.97
Product Information
  • Catalog #
    Length
    Width
    Availability
    Price
  • Paper fasteners
    Paper fasteners
    470149-676
    1"
    1"
    Log in for availability
    Paquete de 100 - $103.20