Orders LinkContactUsLinkComponent
Utility Pails
Utility Pail with Lid, 7.5 L (2 gal.)
  470006-566
 :   Avantor
Utility Pails
  470006-566
 :  Avantor
Utility Pails
Utility Pail with Lid, 7.5 L (2 gal.)
$239.40
Each (1UN)
PE utility pails with lid.
  • Capacity:
    7.5 L (2 gal.)
  • Description:
    Utility Pail with Lid
Accessories
  •  
    Description
    Dimensions
  • 470149-944
    Tagboard
    23×30.5 cm (9×12")
    Paquete de 1 - $176.97
  •  
    Description
    Length
    Width
  • 470149-676
    Paper fasteners
    1"
    1"
    Paquete de 100 - $103.20