Orders LinkContactUsLinkComponent
Graduated Cylinders with Plastic Base
25 ml, 0.5 ml
Catalog # 470188-952
 :  CY137-25
Graduated Cylinders with Plastic Base
Catalog # 470188-952
Graduated Cylinders with Plastic Base
Graduated Cylinders with Plastic Base
25 ml, 0.5 ml
Price
$162.30
Log in Para disponibilidad
Each (1UN) Paquete de 10 (1UN)
Product Details & Documents
Durable Borosilicate Glass Construction.
  • Graduations permanently fused into glass
  • Hexagonal base and bumper guard add protection for all sizes
Double metric scale, calibrated to contain. Borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing.
Especificaciones
  • Graduation Interval:
    0.5 ml
  • Capacity:
    25 ml
  • Cat. No.:
    470188-952
  • Form:
    Tall