Orders LinkContactUsLinkComponent
Banana Plug Patch Cords
Banana Plug Patch Cords
Patch cord, red, 15 cm, banana plugs both ends
Catalog # 470188-742
Supplier #:  1350302190002
Banana Plug Patch Cords
Catalog # 470188-742
Banana Plug Patch Cords
Banana Plug Patch Cords
Patch cord, red, 15 cm, banana plugs both ends
Price
$52.56
Log in for availability
Each (1UN) Paquete de 6 (1UN)
Product Details & Documents
Double ended banana plug jumper wires.
  • Available in red or black
  • 15 cm (6") or 61 cm (24") long
Stackable 4 mm banana plugs on both ends of these flexible, plastic coated wires. The wires have a rating of 15 A, AC.
Specifications
  • Description:
    Patch cord, red, 15 cm, banana plugs both ends