Orders LinkContactUsLinkComponent
Jars and Lids
Jars and Lids
Lid, Metal, 63 mm,for 20469 and 29480, Metal, White
Catalog # 470188-432
Supplier:   EMPIRE/EMCO
Número de proveedor #:  87A001C63A
Jars and Lids
Catalog # 470188-432
Supplier:  EMPIRE/EMCO
Jars and Lids
Jars and Lids
Lid, Metal, 63 mm,for 20469 and 29480, Metal, White
Price
$11.29
Log in Para disponibilidad
Each (1UN) Paquete de 12 (1UN)
Product Details & Documents
Stock up on classroom essentials for every classroom activity.
Especificaciones
  • Color:
    White
  • Description:
    Lid, Metal, 63 mm,for 20469 and 29480
  • Material:
    Metal
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • Screw Cap for Specimen Jar, 80 mm
    Screw Cap for Specimen Jar, 80 mm
    470175-872
    Log in Para disponibilidad
    Each (1UN) - $26.33