Jars and Lids
Jar, Plastic, Wide-Mouth,Tall,12 oz. (350 mL, 63 mm Diameter), Clear, Polystyrene, Clear
Catalog # 470188-430
Supplier: PLASTIC BOTTLES INC.
Número de proveedor #: 354
Jump to:
Product Details & Documents
Stock up on classroom essentials for every classroom activity.
Especificaciones
- Color:Clear
- Description:Jar, Plastic, Wide-Mouth,Tall,12 oz. (350 mL, 63 mm Diameter), Clear
- Material:Polystyrene
Accessories
Información del producto
- Catalog #AvailabilityPrice
- Screw Cap for Specimen Jar, 80 mm