Simple Squamous Epithelium Slides
Simple Squamous Epithelium, Section, Hematoxylin and Eosin
Catalog # 470182-600
Supplier: Avantor
Jump to:
Product Details & Documents
Single layer of epithelium that helps with diffusion and filtration.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Choose from several available stains and preparations for these tissue sections to get the view you're looking for. Slides are made from mammalian kidney tissue.
Hematoxylin and Eosin-stained slides show Bowman's Capsule, the thin section slide is plastic-embedded for greater viewing clarity.
Silver Nitrate-stained slides show Pavement tissue of mesentery, with cell borders in sharp relief.
Especificaciones
- Description:Simple Squamous Epithelium
- Stain:Hematoxylin and Eosin
- Orientation:Section