Orders LinkContactUsLinkComponent
Ward's® Pine Structure Microscope Slides
Ward's® Pine Structure Microscope Slides
Mature embryo, Longitudinal section, Quadruple stained
  470181-212
 :   Avantor
Ward's® Pine Structure Microscope Slides
  470181-212
 :  Avantor
Ward's® Pine Structure Microscope Slides
Ward's® Pine Structure Microscope Slides
Mature embryo, Longitudinal section, Quadruple stained
$154.65
Each (1UN)
Anatomy of a gymnosperm.
  • All aspects of pine live history represented
  • Prepared by expert technicians
Explore all aspects of the gymnosperm life cycle. Pinus is a common educational representative of the gymnosperms exhibiting the general characteristics of the clade. Root, stem, leaf, and reproductive structures are all available.
  • Description:
    Mature embryo
  • Stain:
    Quadruple stained
  • Orientation:
    Longitudinal section