Orders LinkContactUsLinkComponent
Ward's® Lichen Slides
Ward's® Lichen Slides
Crustose, Section, Quadruple
Catalog # 470181-106
Supplier:   Avantor
Ward's® Lichen Slides
Catalog # 470181-106
Supplier:  Avantor
Ward's® Lichen Slides
Ward's® Lichen Slides
Crustose, Section, Quadruple
Price
$381.60
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
A composite of fungi and algae.
  • Ward's produced slides
  • Rigorous quality control standards
A great example of mutualism, lichen are a composite organism of fungi and algae. In these slides both parts of this relationship are easily seen.
Especificaciones
  • Description:
    Crustose
  • Stain:
    Quadruple
  • Orientation:
    Section