Ward's® Lichen Slides
Crustose, Section, Quadruple
Catalog # 470181-106
Supplier: Avantor
Jump to:
Product Details & Documents
A composite of fungi and algae.
Ward's produced slides
- Rigorous quality control standards
A great example of mutualism, lichen are a composite organism of fungi and algae. In these slides both parts of this relationship are easily seen.
Especificaciones
- Description:Crustose
- Stain:Quadruple
- Orientation:Section