Orders LinkContactUsLinkComponent
Ward's® Lichen Slides
Ward's® Lichen Slides
Mycobiont, Whole mount, Fast Green and Mallory's
Catalog # 470181-102
Supplier:   Avantor
Ward's® Lichen Slides
Catalog # 470181-102
Supplier:  Avantor
Ward's® Lichen Slides
Ward's® Lichen Slides
Mycobiont, Whole mount, Fast Green and Mallory's
Price
$182.70
Log in for availability
Each (1UN)
Product Details & Documents
A composite of fungi and algae.
  • Ward's produced slides
  • Rigorous quality control standards
A great example of mutualism, lichen are a composite organism of fungi and algae. In these slides both parts of this relationship are easily seen.
Specifications
  • Description:
    Mycobiont
  • Stain:
    Fast Green and Mallory's
  • Orientation:
    Whole mount