Ward's® Specimen Trays
Ward's® Specimen Trays, 8", ¹¹⁄₁₆", 6"
470178-314
: 306140
Make use of every inch of drawer space in your cabinets by combining these differently-sized Specimen Trays.
Made of strong pasteboard, the trays are finished outside in black gloss paper and lined with white. Measurements reflect outside dimensions.
- Width:8"
- Description:Ward's® Specimen Trays
- Height:¹¹⁄₁₆"
- Length:6"