Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Ward's® Specimen Trays

Ward's® Specimen Trays, 3", ¹¹⁄₁₆", 2"
Catalog # 470178-308
Supplier:   CHARLES W. KANTY
 :  306110
Ward's® Specimen Trays
Catalog # 470178-308
Supplier:  CHARLES W. KANTY
Ward's® Specimen Trays
Ward's® Specimen Trays

Ward's® Specimen Trays, 3", ¹¹⁄₁₆", 2"

Price
$40.31
Log in Para disponibilidad
Each (1UN) Paquete de 100 (1UN) Case de 1,000 (1UN)
Product Details & Documents

Make use of every inch of drawer space in your cabinets by combining these differently-sized Specimen Trays.

Made of strong pasteboard, the trays are finished outside in black gloss paper and lined with white. Measurements reflect outside dimensions.

Especificaciones
  • Width:
    3"
  • Description:
    Ward's® Specimen Trays
  • Height:
    ¹¹⁄₁₆"
  • Length:
    2"