Ward's® Introduction To Microscopes Slides
Computer chip, Whole mount, Unstained
470177-672
: Avantor
Examine how a microscope works.
- Professionally made microscope slides
- Explore basic microscope functions and environmental materials
Introduce students to the world of microscopy and the world around them with these prepared slides. Use a selection of text slides to show students how microscopes change how you view materials. Students can then examine items that exist in the world around them.
- Description:Computer chip
- Stain:Unstained
- Orientation:Whole mount