Stirring Rods
Stirring Rod, Glass, 5 mm x 200 mm (8")
Catalog # 470176-070
Jump to:
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
STIRRING ROD GLASS 200MM L 5MM D PK12 Catalog #470020-788
Product 470020-788 is similar to 470176-070 but manufacturer and specifications may vary.
STIRRING ROD GLS 200X5MM PK100 Catalog #59060-069
Product 59060-069 is similar to 470176-070 but manufacturer and specifications may vary.
Product Details & Documents
Annealed flint glass rods with fire polished ends.
- Both ends fire polished and rounded
Stirring rods are made of soft flint glass, with both ends fire-polished and rounded.
Especificaciones
- Description:Stirring Rod, Glass, 5 mm x 200 mm (8")