Orders LinkContactUsLinkComponent
Alloy Tuning Fork Sets
Alloy Tuning Fork Sets
Tuning Forks, Set of 8
Catalog # 470104-594
Supplier:   Avantor
Alloy Tuning Fork Sets
Catalog # 470104-594
Supplier:  Avantor
Alloy Tuning Fork Sets
Alloy Tuning Fork Sets
Tuning Forks, Set of 8
Price
$1,527.48
Log in for availability
Kit de 8
Product Details & Documents
Two sets of popular tuning forks are available.
  • Useful Selection of Frequencies
  • Two Sets Available
  • Setting from A to G
One set features four tuning forks (256 C, 320 E, 384 G, 512C).
The other set features eight tuning forks (256 C, 320 E, 384 G, 480 B, 288 D, 341.3 F, 426.7 A, 512 C).
Specifications
  • Description:
    Tuning Forks, Set of 8