Orders LinkContactUsLinkComponent
Boreal Standard Compound Microscopes
Boreal Standard Compound Microscopes
Compound microscope, Abbe, LED corded, Mechanical
  470177-238
Supplier #:  5584500
Boreal Standard Compound Microscopes
  470177-238
Boreal Standard Compound Microscopes
Boreal Standard Compound Microscopes
Compound microscope, Abbe, LED corded, Mechanical
$8,378.87
Each (1UN)
Boreal standard microscopes feature metal construction, a rugged student-proof design, and precision optics making them a favorite among educators.
  • Coated-glass optics
  • Wide-field eyepiece with pointer
  • Limited lifetime warranty
These microscopes optics are parfocal and parcentered for a crisp, consistent field of view. All units feature strategically located controls, course and fine focus, and a 360° rotatable head. Units are available in a variety of configurations offering three or four objectives, plain or mechanical stage, and corded or cordless charging.
Ordering information: Microscopes include a manual, dust cover, and limited warranty.
  • Description:
    Compound microscope
  • Stage:
    Mechanical
  • Condenser Type:
    Abbe
  • Illumination:
    LED corded
  • Cat. No.:
    470177-238
  • Objectives:
    4×, 10×, 40×, 100×
Accessories
  •  
    Description
    Electrical
  • 470005-050
    Replacement bulb
    1 W
    Each (1UN) - $159.92
  •  
    Description
    Electrical
  • 470021-694
    Replacement Bulb, with Bayonet Base
    110 V, 20 W
    Each (1UN) - $133.26