Nostoc Slide
Nostoc, Section, Safranin
Catalog # 470176-976
Supplier: Avantor
Jump to:
Product Details & Documents
A genus of cyanobacteria.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Filaments and heterocysts with occasional spores, within sheath.
Especificaciones
- Description:Nostoc
- Stain:Safranin
- Orientation:Section