Orders LinkContactUsLinkComponent
Nostoc Slide
Nostoc Slide, Whole Mount, Safranin
  470176-842
 :   Avantor
<i>Nostoc</i> Slide
  470176-842
 :  Avantor
<i>Nostoc</i> Slide
Nostoc Slide
Nostoc Slide, Whole Mount, Safranin
$148.35
Each (1UN)
A genus of cyanobacteria.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Filaments and heterocysts with occasional spores, within sheath.
  • Description:
    Nostoc Slide
  • Stain:
    Safranin
  • Orientation:
    Whole Mount