Tri Pour Beakers
50 ml (1.7 oz.), Tri-pour beakers
470174-588
: 80092
These unbreakable polypropylene beakers feature a tri-cornered lip for dripless pouring.
Accurate graduations to ±5
- Tri-cornered lip for dripless pouring
- Unbreakable
- Resistant to common acids, solvents, and alkalis
- Autoclavable
They are unaffected by commonly used acids, alkalis, and solvents and are autoclavable.
Graduations are accurate to ±5.
- Capacity:50 ml (1.7 oz.)
- Description:Tri-pour beakers