Orders LinkContactUsLinkComponent
Tri Pour Beakers
100 ml (3.4 oz.), Tri-pour beakers
  470174-586
 :  80093
Tri Pour Beakers
  470174-586
 :  
Tri Pour Beakers
Tri Pour Beakers
100 ml (3.4 oz.), Tri-pour beakers
These unbreakable polypropylene beakers feature a tri-cornered lip for dripless pouring.
  • Accurate graduations to ±5
  • Tri-cornered lip for dripless pouring
  • Unbreakable
  • Resistant to common acids, solvents, and alkalis
  • Autoclavable
They are unaffected by commonly used acids, alkalis, and solvents and are autoclavable.
Graduations are accurate to ±5.
  • Capacity:
    100 ml (3.4 oz.)
  • Description:
    Tri-pour beakers