Tri Pour Beakers
100 ml (3.4 oz.), Tri-pour beakers
470174-586
: 80093
These unbreakable polypropylene beakers feature a tri-cornered lip for dripless pouring.
Accurate graduations to ±5
- Tri-cornered lip for dripless pouring
- Unbreakable
- Resistant to common acids, solvents, and alkalis
- Autoclavable
They are unaffected by commonly used acids, alkalis, and solvents and are autoclavable.
Graduations are accurate to ±5.
- Capacity:100 ml (3.4 oz.)
- Description:Tri-pour beakers