Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FRASCO NUNCLON 260ML TISS CULT PK5
Catalog # 470174-472
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
FRASCO NUNCLON 260ML TISS CULT PK5
Catalog # 470174-472
Supplier:
Thermo Fisher Scientific
Supplier Number:
156472
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
470174-472_162134
SC15708-124_160806
SC15708-124_161854
SC15708-124_163389
Product Literature
Thermo Scientific Nunc Cell Culture Plastics Guide
Documentation
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International