Culture Flasks
50 mL
: 163371
Jump to:
Discontinued
Product Details & Documents
Specially Coated Tissue Culture Flasks.
- Nunclon surface treatment promotes cell attachment and growth
- Easy filling and access for pipets
- Stackable, space saving design
Cell adherence is discouraged from the untreated sidewalls, neck and top panel.
Especificaciones
- Description:50 mL