Disposable Polyethylene Dropper Pipettes
470161-252
These polyethylene pipettes are chemically inert.
- For consistent bulb pressure and top circumference
- Disposable
Ensure precisely measured drop reproducibility.
These polyethylene pipettes are chemically inert.
Ensure precisely measured drop reproducibility.