Orders LinkContactUsLinkComponent
Disposable Polyethylene Dropper Pipettes
  470161-252
Disposable Polyethylene Dropper Pipettes
  470161-252
 :  
Disposable Polyethylene Dropper Pipettes
Disposable Polyethylene Dropper Pipettes
These polyethylene pipettes are chemically inert.
  • For consistent bulb pressure and top circumference
  • Disposable
Ensure precisely measured drop reproducibility.