Orders LinkContactUsLinkComponent
Ward's® Igneous Rock Collection
Igneous Rock Collection, Hand Specimen Set, 12
  470154-094
 :   Avantor
Ward's® Igneous Rock Collection
  470154-094
 :  Avantor
Ward's® Igneous Rock Collection
Ward's® Igneous Rock Collection
Igneous Rock Collection, Hand Specimen Set, 12
$2,964.48
Each (1UN)
Samples of Intrusive and Extrusive Igneous Rocks
  • Introduce concepts of the Earth's crust
  • Compartmented box
Provide your students with an excellent introduction to the forces at work beneath the Earth’s crust. The set is housed in a compartmented collection box and includes an identification list.
Contents:
12 numbered samples, 1" x 2"
Rocks:
basalt, diorite, gabbro, granite, obsidian, pegmatite, peridotite, pumice, rhyolite (2), scoria, syenite
Hand Specimen Set:
Large hand samples are ideal for demonstration and display. This set is housed in individual, labeled plastic display trays and comes with an identification list.
Contents:
12 numbered samples, 3" x 4"
  • Description:
    Igneous Rock Collection, Hand Specimen Set
  • No. of Samples:
    12