Orders LinkContactUsLinkComponent
Sandpaper
Sandpaper
Medium grit, 2×2"
Catalog # 470152-956
Supplier:   Avantor
Sandpaper
Catalog # 470152-956
Supplier:  Avantor
Sandpaper
Medium grit, 2×2"
Price
$39.50
Log in for availability
Each (1UN)
Product Details & Documents
Medium grade, 180 grit. 9×11" sheets.
Specifications
  • Description:
    Medium grit
  • Dimensions:
    2×2"