Waterproof Field Notebooks
Waterproof Field Notebooks, Shirt-Pocket; 50 Sheets, 3", 5"
470152-784
: 135
An absolute must when conducting field studies.
- Take legible notes in any conditions
- Easy to spot if accidentally dropped
- Aligned with GLOBE protocols
Unique all–weather paper lets you take and keep legible notes regardless of the conditions, and the bright yellow cover is easy to spot if accidentally dropped. The field version has one page for data values and on the facing page it has a grid for drawing.
- Description:Waterproof Field Notebooks, Shirt-Pocket; 50 Sheets
- Length:5"
- Width:3"



