Orders LinkContactUsLinkComponent
Latex Disposable Gloves
Powdered Latex, L, White, 100 Gloves/Box
Catalog # 470152-194
 :  R10UN-LG
Latex Disposable Gloves
Catalog # 470152-194
Latex Disposable Gloves
Latex Disposable Gloves
Powdered Latex, L, White, 100 Gloves/Box
Price
$677.17
Log in for availability
Caja de 100 Case de 10 (100UN)
Product Details & Documents
With excellent tactile sensitive, these wrist-length gloves are ambidextrous and disposable.
  • Ambidextrous
  • Non sterile
  • Economy hand protection
  • Wrist-length
  • Aligned with GLOBE protocols
Perfect for student use, they feature a textured finish for sure grip and a beaded cuff to keep them in place.
Latex gloves reduce costs for hand protection when glove usage is high. Pre-powered option available for easy removal in your choice of sizes.
Specifications
  • Glove Color:
    White
  • Glove size:
    L
  • Description:
    Powdered Latex
  • Packaging:
    100 Gloves/Box
  • Cat. No.:
    470152-194
  • Latex / latex-free:
    Latex
  • Disposable / reusable:
    Disposable
  • Ambidextrous / hand-specific:
    Ambidextrous
Accessories
Product Information
  • Catalog #
    Glove size
    Glove Color
    Availability
    Price
  • Latex Gloves
    Latex Gloves
    470150-774
    XL
    White
    Log in for availability
    Paquete de 100 - $671.57