Latex Disposable Gloves
Powdered Latex, L, White, 100 Gloves/Box
Catalog # 470152-194
Supplier: DIVAL SAFETY & SUPPLIES SE
: R10UN-LG
Jump to:
Product Details & Documents
With excellent tactile sensitive, these wrist-length gloves are ambidextrous and disposable.
- Ambidextrous
- Non sterile
- Economy hand protection
- Wrist-length
- Aligned with GLOBE protocols
Perfect for student use, they feature a textured finish for sure grip and a beaded cuff to keep them in place.
Latex gloves reduce costs for hand protection when glove usage is high. Pre-powered option available for easy removal in your choice of sizes.
Specifications
- Glove Color:White
- Glove size:L
- Description:Powdered Latex
- Packaging:100 Gloves/Box
- Cat. No.:470152-194
- Latex / latex-free:Latex
- Disposable / reusable:Disposable
- Ambidextrous / hand-specific:Ambidextrous
Accessories
Product Information
- Catalog #Glove sizeGlove ColorAvailabilityPrice
- Latex Gloves