Precision pH Test Strips
pH paper strips, 3.0–6.0
Catalog # 470150-846
Supplier: PRECISION LABORATORIES SE
Número de proveedor #: PH3060-1B-50
Jump to:
Product Details & Documents
The indicator on these strips won’t contaminate the solutions the strips are testing.
- Vinyl-backed
- Non-contaminating indicator
- Fifty strips per package
Durable pH testing strips that are backed with vinyl.
Especificaciones
- Description:pH paper strips
- pH Range:3.0–6.0