Order Entry
Mexico
Orders LinkContactUsLinkComponent
United Scientific™ Push/Pull Spring Scales

United Scientific™ Push/Pull Spring Scales

Push/pull spring scale, brown, 1000 g/10 N, graduation interval: 0.2 N
Catalog # 470150-486
Supplier #:  PS1000
United Scientific™ Push/Pull Spring Scales
Catalog # 470150-486
United Scientific™ Push/Pull Spring Scales
United Scientific™ Push/Pull Spring Scales

Push/pull spring scale, brown, 1000 g/10 N, graduation interval: 0.2 N

Price
$142.53
Log in for availability
Each (1UN) Case de 150 (1UN)
Product Details & Documents

Versatile spring scale.

  • Weighs in Grams and Newtons
  • Color coded to differentiate capacity

These Push Pull Spring Scales are a versatile spring scale that can be used to measure forces using push or pull functions, or to weigh a suspended object from the large hook.

Delivery information: Includes a zero adjustment screw, a metal suspension ring, instructions and application sheets.

Caution: This product is for ages six and up.

Specifications
  • Description:
    Push/pull spring scale, brown
  • Weighing Capacity:
    1000 g/10 N, graduation interval: 0.2 N