Enameled Copper Magnet Wire
Wire, Enameled Copper Magnet, 20 Gauge, 94 m, 454 grams
Catalog # 470149-670
Supplier: ARCOR ELECTRONICS CO INC SE
Número de proveedor #: E-20
Jump to:
Product Details & Documents
A high quality enameled, copper magnet wire.
The 454 g spools are available in several standard AWG sizes.
Especificaciones
- Description:Wire, Enameled Copper Magnet, 20 Gauge, 94 m, 454 grams
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



